Assay Details
| Gene Symbol: SLCO1B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315607.1 | NP_001302536.1 | 8 - 9 | 1131 | 88 | |
| XM_005655659.3 | XP_005655716.1 | 1259 | 88 | |||
| XM_021091087.1 | XP_020946746.1 | 1293 | 88 | |||
| XM_021091089.1 | XP_020946748.1 | 1425 | 88 | |||
| XM_021091090.1 | XP_020946749.1 | 1430 | 88 | |||
| XM_021091091.1 | XP_020946750.1 | 1115 | 88 | |||
| GenBank mRNA | LN833582.1 | - | 8 - 9 | 1131 | 88 | |
More Information
Set Membership: |
|

SLCO1B3
Genomic Map