Assay Details
| Gene Symbol: NMUR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315615.1 | NP_001302544.1 | 2 - 3 | 807 | 100 | |
| GenBank mRNA | KJ622335.1 | - | 2 - 3 | 807 | 100 | |
| KM097067.1 | - | 2 - 3 | 636 | 100 | ||
More Information
Set Membership: |
|

NMUR2
Genomic Map