Assay Details
| Gene Symbol: GRPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315640.1 | NP_001302569.1 | 1 - 2 | 564 | 103 | |
| GenBank mRNA | AK231328.1 | - | 1 - 2 | 810 | 103 | |
| KM052179.1 | - | 1 - 2 | 564 | 103 | ||
More Information
Set Membership: |
|

GRPR
Genomic Map