Assay Details
| Gene Symbol: ADH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315670.1 | NP_001302599.1 | 260 | 113 | ||
| GenBank mRNA | KP735778.1 | - | 260 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001315670.1 transcripts.
Set Membership: |
|

ADH4
Genomic Map