Assay Details
| Gene Symbol: EBI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315682.1 | NP_001302611.1 | 4 - 5 | 558 | 84 | |
| XM_005661336.3 | XP_005661393.1 | 1405 | 84 | |||
| GenBank mRNA | AB126827.1 | - | 4 - 5 | 559 | 84 | |
More Information
Set Membership: |
|

EBI3
Genomic Map