Assay Details
| Gene Symbol: LOC100738724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315744.1 | NP_001302673.1 | 7 - 8 | 1153 | 70 | |
| Gene Symbol: CYP39A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | KP687248.1 | - | 7 - 8 | 1153 | 70 | |
More Information
Set Membership: |
|

LOC100738724
Genomic Map