Assay Details
| Gene Symbol: CLEC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315794.1 | NP_001302723.1 | 2 - 3 | 156 | 81 | |
| GenBank mRNA | KJ786417.1 | - | 2 - 3 | 156 | 81 | |
More Information
Set Membership: |
|

CLEC1B
Genomic Map