Assay Details
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348770.1 | NP_001335699.1 | 1 - 2 | 83 | 95 | |
| NR_145981.2 | 1 - 2 | 83 | 95 | |||
| GenBank mRNA | AB116559.1 | - | 1 - 2 | 42 | 95 | |
| AB635379.1 | - | 1 - 2 | 45 | 95 | ||
| AK238831.1 | - | 1 - 2 | 66 | 95 | ||
| AK239664.1 | - | 1 - 2 | 64 | 95 | ||
| AK398365.1 | - | 1 - 2 | 72 | 95 | ||
| AY438642.1 | - | 1 - 2 | 61 | 95 | ||
More Information
Set Membership: |
|

CD8B
Genomic Map