Assay Details
| Gene Symbol: AKT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159776.1 | NP_001153248.1 | 2 | 1400 | 61 | |
| XM_021081499.1 | XP_020937158.1 | 1587 | 61 | |||
| XM_021081500.1 | XP_020937159.1 | 1560 | 61 | |||
| XM_021081501.1 | XP_020937160.1 | 1844 | 61 | |||
| GenBank mRNA | AB499527.1 | - | 2 | 1400 | 61 | |
| HQ687753.1 | - | 2 | 1380 | 61 | ||
More Information
Set Membership: |
|

AKT1
Genomic Map