Assay Details
| Gene Symbol: CLCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214148.3 | NP_999313.2 | 10 - 11 | 1721 | 63 | |
| GenBank mRNA | AK345006.1 | - | 10 - 11 | 1723 | 63 | |
More Information
Set Membership: |
|

CLCA1
Genomic Map