Assay Details
| Gene Symbol: IFIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204395.2 | NP_001191324.1 | 1 - 2 | 113 | 112 | |
| GenBank mRNA | AK230808.1 | - | 1 - 2 | 70 | 112 | |
| AK230895.1 | - | 1 - 2 | 70 | 112 | ||
| AK343662.1 | - | 1 - 2 | 70 | 112 | ||
| AK389734.1 | - | 1 - 2 | 113 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001204395.2 transcripts.
Set Membership: |
|

IFIT3
Genomic Map