Assay Details
| Gene Symbol: PRIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243669.1 | NP_001230598.1 | 7 - 8 | 830 | 88 | |
| XM_021090961.1 | XP_020946620.1 | 956 | 88 | |||
| GenBank mRNA | AK344370.1 | - | 7 - 8 | 830 | 88 | |
More Information
Set Membership: |
|

PRIM1
Genomic Map