Assay Details
| Gene Symbol: MALL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243633.1 | NP_001230562.1 | 1 | 121 | 118 | |
| XM_021085752.1 | XP_020941411.1 | 2276 | 118 | |||
| GenBank mRNA | AK231533.1 | - | 1 | 121 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243633.1, XM_021085752.1 transcripts.
Set Membership: |
|

MALL
Genomic Map