Assay Details
| Gene Symbol: LOC100622246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315746.1 | NP_001302675.1 | 3 | 823 | 103 | |
| GenBank mRNA | AK236326.1 | - | 3 | 923 | 103 | |
| AK238510.1 | - | 3 | 807 | 103 | ||
| AK351374.1 | - | 3 | 837 | 103 | ||
| AK394847.1 | - | 3 | 769 | 103 | ||
| AK397139.1 | - | 3 | 796 | 103 | ||
| AK398160.1 | - | 3 | 841 | 103 | ||
| AK398199.1 | - | 3 | 744 | 103 | ||
| AK398280.1 | - | 3 | 818 | 103 | ||
| AK400785.1 | - | 3 | 841 | 103 | ||
| AK400979.1 | - | 3 | 719 | 103 | ||
| AK400981.1 | - | 3 | 844 | 103 | ||
| KP735789.1 | - | 3 | 823 | 103 | ||
More Information
Set Membership: |
|

LOC100622246
Genomic Map