Assay Details
| Gene Symbol: ZDHHC21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063619.1 | XP_020919278.1 | 696 | 86 | ||
| XM_021063627.1 | XP_020919286.1 | 732 | 86 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZDHHC21| Gene Symbol: ZDHHC21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063619.1 | XP_020919278.1 | 696 | 86 | ||
| XM_021063627.1 | XP_020919286.1 | 732 | 86 | |||
Set Membership: |
|