Assay Details
| Gene Symbol: PDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927439.5 | XP_001927474.1 | 2337 | 64 | ||
| XM_013986015.2 | XP_013841469.1 | 2491 | 64 | |||
| XM_021080536.1 | XP_020936195.1 | 2430 | 64 | |||
| XM_021080537.1 | XP_020936196.1 | 2590 | 64 | |||
| XM_021080538.1 | XP_020936197.1 | 1352 | 64 | |||
| GenBank mRNA | AB529867.1 | - | 3 - 4 | 236 | 64 | |
| AK233858.1 | - | 3 - 4 | 261 | 64 | ||
More Information
Set Membership: |
|

PDK3
Genomic Map