Assay Details
| Gene Symbol: ARG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928679.5 | XP_001928714.1 | 485 | 78 | ||
| XM_013989111.2 | XP_013844565.1 | 294 | 78 | |||
| GenBank mRNA | AK234497.1 | - | 3 - 4 | 429 | 78 | |
| AK348565.1 | - | 3 - 4 | 429 | 78 | ||
| AK389895.1 | - | 3 - 4 | 429 | 78 | ||
More Information
Set Membership: |
|

ARG2
Genomic Map