Assay Details
| Gene Symbol: LOC102160336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021071421.1 | XP_020927080.1 | 426 | 85 | ||
| GenBank mRNA | AK347921.1 | - | 3 - 4 | 369 | 85 | |
More Information
Set Membership: |
|

LOC102160336
Genomic Map