Assay Details
| Gene Symbol: UBE2R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064700.1 | XP_020920359.1 | 786 | 70 | ||
| GenBank mRNA | AK348709.1 | - | 1 - 2 | 499 | 70 | |
More Information
Set Membership: |
|

UBE2R2
Genomic Map