Assay Details
| Gene Symbol: PDPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093788.1 | XP_020949447.1 | 2176 | 59 | ||
| XM_021093789.1 | XP_020949448.1 | 2384 | 59 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PDPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093788.1 | XP_020949447.1 | 2176 | 59 | ||
| XM_021093789.1 | XP_020949448.1 | 2384 | 59 | |||
Set Membership: |
|