Assay Details
| Gene Symbol: BTNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482157.3 | XP_003482205.2 | 1557 | 59 | ||
| XM_013977699.2 | XP_013833153.1 | 1497 | 59 | |||
| XM_013977700.2 | XP_013833154.1 | 1533 | 59 | |||
| XM_013977701.2 | XP_013833155.1 | 1503 | 59 | |||
| XM_021098499.1 | XP_020954158.1 | 1443 | 59 | |||
| XM_021098500.1 | XP_020954159.1 | 1479 | 59 | |||
More Information
Set Membership: |
|

BTNL2
Genomic Map