Assay Details
| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005667914.3 | XP_005667971.1 | 1077 | 68 | ||
| XM_013980016.2 | XP_013835470.1 | 2241 | 68 | |||
| XM_013980017.2 | XP_013835471.1 | 999 | 68 | |||
More Information
Set Membership: |
|

BRINP3
Genomic Map