Assay Details
| Gene Symbol: PLD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005667950.3 | XP_005668007.1 | 268 | 68 | ||
| XM_005667951.3 | XP_005668008.2 | 362 | 68 | |||
| XM_021064453.1 | XP_020920112.1 | 1407 | 68 | |||
| XM_021064454.1 | XP_020920113.1 | 1428 | 68 | |||
| XM_021064455.1 | XP_020920114.1 | 541 | 68 | |||
| XM_021064456.1 | XP_020920115.1 | 1451 | 68 | |||
More Information
Set Membership: |
|

PLD5
Genomic Map