Assay Details
Gene Symbol: SEBOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005669098.3 | XP_005669155.2 | 788 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
