Assay Details
| Gene Symbol: GMNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005670072.3 | XP_005670129.1 | 956 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005670072.3 transcripts.
Set Membership: |
|

GMNC
Genomic Map