Assay Details
| Gene Symbol: CCDC110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005671722.3 | XP_005671779.1 | 433 | 87 | ||
| XM_021075584.1 | XP_020931243.1 | 328 | 87 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC110| Gene Symbol: CCDC110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005671722.3 | XP_005671779.1 | 433 | 87 | ||
| XM_021075584.1 | XP_020931243.1 | 328 | 87 | |||
Set Membership: |
|