Assay Details
| Gene Symbol: COL21A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013977769.1 | XP_013833223.1 | 3038 | 65 | ||
| XM_021098588.1 | XP_020954247.1 | 1317 | 65 | |||
| XM_021098589.1 | XP_020954248.1 | 822 | 65 | |||
More Information
Set Membership: |
|

COL21A1
Genomic Map