Assay Details
| Gene Symbol: MSANTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013978508.2 | XP_013833962.2 | 1199 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MSANTD1| Gene Symbol: MSANTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013978508.2 | XP_013833962.2 | 1199 | 93 | ||