Assay Details
| Gene Symbol: LOC100525933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013978753.2 | XP_013834207.2 | 205 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013978753.2 transcripts.
Set Membership: |
|

LOC100525933
Genomic Map