Assay Details
| Gene Symbol: ITPRID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013985733.2 | XP_013841187.1 | 748 | 76 | ||
| XM_021079198.1 | XP_020934857.1 | 1078 | 76 | |||
| XM_021079200.1 | XP_020934859.1 | 1078 | 76 | |||
| XM_021079201.1 | XP_020934860.1 | 1078 | 76 | |||
| XM_021079202.1 | XP_020934861.1 | 1078 | 76 | |||
| XM_021079203.1 | XP_020934862.1 | 716 | 76 | |||
| XM_021079204.1 | XP_020934863.1 | 557 | 76 | |||

ITPRID1
Genomic Map