Assay Details
| Gene Symbol: LMX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013988077.2 | XP_013843531.2 | 3713 | 60 | ||
| XM_013988078.2 | XP_013843532.2 | 3641 | 60 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LMX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013988077.2 | XP_013843531.2 | 3713 | 60 | ||
| XM_013988078.2 | XP_013843532.2 | 3641 | 60 | |||
Set Membership: |
|