Assay Details
Gene Symbol: RALYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013996648.2 | XP_013852102.2 | 1435 | 91 | ||
XM_021090619.1 | XP_020946278.1 | 8624 | 91 | |||
XM_021090620.1 | XP_020946279.1 | 1690 | 91 | |||
XM_021090621.1 | XP_020946280.1 | 8753 | 91 | |||
XM_021090622.1 | XP_020946281.1 | 8732 | 91 | |||
XM_021090623.1 | XP_020946282.1 | 714 | 91 | |||
XM_021090624.1 | XP_020946283.1 | 531 | 91 | |||
XM_021090625.1 | XP_020946284.1 | 1819 | 91 | |||
XM_021090626.1 | XP_020946285.1 | 1745 | 91 | |||
XM_021090627.1 | XP_020946286.1 | 2410 | 91 | |||
XM_021090628.1 | XP_020946287.1 | 1576 | 91 | |||
XM_021090629.1 | XP_020946288.1 | 610 | 91 | |||
XM_021090630.1 | XP_020946289.1 | 889 | 91 | |||
XM_021090631.1 | XP_020946290.1 | 739 | 91 | |||
XM_021090632.1 | XP_020946291.1 | 6434 | 91 | |||
XM_021090633.1 | XP_020946292.1 | 1853 | 91 | |||
XM_021090634.1 | XP_020946293.1 | 656 | 91 | |||
XM_021090635.1 | XP_020946294.1 | 951 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
