Assay Details
| Gene Symbol: CPA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013996693.2 | XP_013852147.2 | 732 | 71 | ||
| XM_021089270.1 | XP_020944929.1 | 1645 | 71 | |||
| XM_021089271.1 | XP_020944930.1 | 775 | 71 | |||
More Information
Set Membership: |
|

CPA6
Genomic Map