Assay Details
Gene Symbol: NLRP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013998823.2 | XP_013854277.2 | 3277 | 106 | ||
XM_021095072.1 | XP_020950731.1 | 3208 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
