Assay Details
Gene Symbol: NLRP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013998823.2 | XP_013854277.2 | 2595 | 91 | ||
XM_021095072.1 | XP_020950731.1 | 2526 | 91 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NLRP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013998823.2 | XP_013854277.2 | 2595 | 91 | ||
XM_021095072.1 | XP_020950731.1 | 2526 | 91 |