Assay Details
Gene Symbol: RBMXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021062312.1 | XP_020917971.1 | 523 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021062312.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
