Assay Details
| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925305.6 | XP_001925340.3 | 728 | 80 | ||
| XM_005671945.3 | XP_005672002.1 | 774 | 80 | |||
| XM_005671948.3 | XP_005672005.1 | 630 | 80 | |||
| XM_005671950.3 | XP_005672007.1 | 878 | 80 | |||
| XM_005671953.3 | XP_005672010.1 | 570 | 80 | |||
| XM_021075353.1 | XP_020931012.1 | 892 | 80 | |||
| XM_021075354.1 | XP_020931013.1 | 794 | 80 | |||
| XM_021075355.1 | XP_020931014.1 | 1042 | 80 | |||
| XM_021075356.1 | XP_020931015.1 | 734 | 80 | |||
More Information
Set Membership: |
|

ERICH2
Genomic Map