Assay Details
| Gene Symbol: MAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121114.4 | XP_003121162.1 | 577 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003121114.4 transcripts.
Set Membership: |
|

MAS1
Genomic Map