Assay Details
Gene Symbol: LOC106509183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021064249.1 | XP_020919908.1 | 717 | 77 | ||
XM_021064260.1 | XP_020919919.1 | 717 | 77 | |||
XM_021064264.1 | XP_020919923.1 | 718 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
