Assay Details
| Gene Symbol: LOC106509183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064249.1 | XP_020919908.1 | 1372 | 68 | ||
| XM_021064260.1 | XP_020919919.1 | 1372 | 68 | |||
| XM_021064264.1 | XP_020919923.1 | 1262 | 68 | |||
More Information
Set Membership: |
|

LOC106509183
Genomic Map