Assay Details
| Gene Symbol: LOC100517051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064287.1 | XP_020919946.1 | 376 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100517051| Gene Symbol: LOC100517051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064287.1 | XP_020919946.1 | 376 | 89 | ||