Assay Details
| Gene Symbol: UCMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064863.1 | XP_020920522.1 | 227 | 68 | ||
| XM_021064864.1 | XP_020920523.1 | 227 | 68 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: UCMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064863.1 | XP_020920522.1 | 227 | 68 | ||
| XM_021064864.1 | XP_020920523.1 | 227 | 68 | |||
Set Membership: |
|