Assay Details
Gene Symbol: SLITRK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021065781.1 | XP_020921440.1 | 161 | 76 | ||
XM_021065782.1 | XP_020921441.1 | 162 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021065781.1, XM_021065782.1 transcripts.
Set Membership: |
![]() ![]() |
