Assay Details
| Gene Symbol: SLITRK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065781.1 | XP_020921440.1 | 198 | 90 | ||
| XM_021065782.1 | XP_020921441.1 | 199 | 90 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLITRK6| Gene Symbol: SLITRK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065781.1 | XP_020921440.1 | 198 | 90 | ||
| XM_021065782.1 | XP_020921441.1 | 199 | 90 | |||
Set Membership: |
|