Assay Details
| Gene Symbol: LOC100624995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066303.1 | XP_020921962.1 | 1793 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100624995| Gene Symbol: LOC100624995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066303.1 | XP_020921962.1 | 1793 | 100 | ||