Assay Details
| Gene Symbol: LOC106505461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067303.1 | XP_020922962.1 | 1271 | 87 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106505461| Gene Symbol: LOC106505461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067303.1 | XP_020922962.1 | 1271 | 87 | ||