Assay Details
| Gene Symbol: LOC100623616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069630.1 | XP_020925289.1 | 697 | 84 | ||
| XM_021069631.1 | XP_020925290.1 | 765 | 84 | |||
More Information
Set Membership: |
|

LOC100623616
Genomic Map