Assay Details
| Gene Symbol: TAFA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070829.1 | XP_020926488.1 | 950 | 125 | ||
| XM_021070831.1 | XP_020926490.1 | 951 | 125 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021070829.1, XM_021070831.1 transcripts.
Set Membership: |
|

TAFA1
Genomic Map