Assay Details
| Gene Symbol: KCNJ6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070975.1 | XP_020926634.1 | 1495 | 101 | ||
| XM_021070976.1 | XP_020926635.1 | 1550 | 101 | |||
| XM_021070977.1 | XP_020926636.1 | 2175 | 101 | |||
| XM_021070978.1 | XP_020926637.1 | 2174 | 101 | |||
More Information
Set Membership: |
|

KCNJ6
Genomic Map