Assay Details
Gene Symbol: GRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021071050.1 | XP_020926709.1 | 1102 | 72 | ||
XM_021071051.1 | XP_020926710.1 | 1102 | 72 | |||
XM_021071052.1 | XP_020926711.1 | 1102 | 72 | |||
XM_021071054.1 | XP_020926713.1 | 1102 | 72 | |||
XM_021071055.1 | XP_020926714.1 | 1102 | 72 | |||
XM_021071056.1 | XP_020926715.1 | 12960 | 72 | |||
XM_021071057.1 | XP_020926716.1 | 12991 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
